Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TcBuster-M Transposase Antibody, Novus Biologicals™

Catalog No. p-200104884
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11511SP 25 μg
MAB11511100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11511SP Supplier R&D Systems Supplier No. MAB11511SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TcBuster Transposase Monoclonal antibody specifically detects TcBuster Transposase in Broad Species Reactivity samples. It is validated for Western Blot,Simple Western

Specifications

Antigen TcBuster Transposase
Applications Western Blot, Simple Western
Classification Monoclonal
Clone 1077035
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Simple Western 20 μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS.
Gene Alias GLEAN_06077, TcasGA2_TC006077, Transposase
Host Species Mouse
Immunogen Synthetic Peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Target Species Broad Species Reactivity
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.