Learn More
Thermo Scientific TCC-3000RS/SD SS VALV POD 6/EA
Supplier:  Thermo Scientific 6722.9013
Encompass
Stainless steel HP, 2-position, 6-port non-biocompatible valve pod for pressures up to 41 MPa (6,000 psi)For Use With (Equipment) TCC-3000 RS/SD Thermostatted Column CompartmentMaterial Stainless SteelMax. Pressure 6000 psi (413 bar)Product Line UltiMateType Valve PodNo. of Ports 6Unit Size Each
Catalog No. NC2320684
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.