Learn More
AdipoGen TCEP (2 g)
SDP
Supplier:  AdipoGen CDXT0176G002
Encompass_Preferred
Chemical. CAS: 51805-45-9. Formula: C9H15O6P·HCl. MW: 286.60. TCEP is an odorless, selective, and water-soluble strong reducing agent that is commonly used in biochemistry and molecular biology applications. It is commonly used to rapidly reduce protein and peptide disulfide bonds a preparatory step for gel electrophoresis. TCEP can be combined with proteases to simultaneously reduce and digest proteins prior to mass spectrometry in order to dramatically increase sequence coverage. More stable than DTT and useful in mass spectrometry applications. It has also been used to measure ascorbic acid and dehydroascorbic acid in biological samples. TCEP is a useful chelating agent for various heavy metal ions as Zn(II), Cd(II), Pb(II), and Ni(II).
Catalog No. 50-205-4660
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.