Learn More
TCF-7/TCF-1 Mouse anti-Mouse, Human, Alexa Fluor 647, Clone: S33-966, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | TCF-7/TCF-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | S33-966 |
| Conjugate | Alexa Fluor 647 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | TCF7; Transcription factor 7; TCF-1; TCF1; T-cell factor 1 |
| Host Species | Mouse |
| Immunogen | Mouse TCF-7 Recombinant Protein |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.