Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human AGA His Protein
SDP

Catalog No. NBP252631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252631 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252631 Supplier Novus Biologicals™ Supplier No. NBP252631
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 24-346 of Human AGA The Recombinant Human AGA Protein is derived from Baculovirus. The Recombinant Human AGA Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 175
Molecular Weight (g/mol) TMW: 35.7kDa
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −70°C long term. Avoid freeze-thaw cycles.
Gene Alias AGU, aspartylglucosaminidase, aspartylglucosylamine deaspartylase, ASRG, EC 3.5.1, EC 3.5.1.26, GA, glycosylasparaginase, N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase, N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase
Common Name AGA
Conjugate Unconjugated
Research Category Cell Cycle and Replication, Proteases & Other Enzymes
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.