Learn More
TCI OPTI-GUARD Fit CH-S, 3/pk , TCI America™
SDP
Catalog No. S38393CLM Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
3pk
Catalog No.
S38393CLM 3pk
1 options
Catalog No. S38393CLM Supplier TCI America Supplier No. S38393CLM
Universal filling for most columns and zero dead-volume connection without any effect on column performance. This series is recommended for the further purification of relatively clean samples using either 4.6 mm I.D. or 2.0 mm I.D. HPLC columns.
Specifications
Length (Metric) 15 mm
Diameter (Metric) 1 mm