Learn More
TCI OPTI-GUARD Fit MB-S, 3/pk , TCI America™
SDP
Catalog No. S38193CLM Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
3pk
Catalog No.
S38193CLM 3pk
1 options
Catalog No. S38193CLM Supplier TCI America Supplier No. S38193CLM
Universal filling for most columns and zero dead-volume connection without any effect on column performance. This series is recommended for the further purification of relatively clean samples using either 4.6 mm I.D. or 2.0 mm I.D. HPLC columns.
Specifications
Length (Metric) 15 mm
Diameter (Metric) 1 mm