Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCLP Pesticides Spiking Mixture, Two Analytes, various concentration in methanol, Ultra Scientific

Catalog No. USTCLP5331
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-TCLP-5331 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-TCLP-5331 Supplier Agilent Technologies Supplier No. TCLP5331
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material TCLP Pesticides Spiking Mixture
CAS Max % 99.24, 0.25, 0.5
CAS 57-74-9, 8001-35-2
Analyte or Component Names Methanol,chlordane,toxaphene
Element Methanol, Toxaphene
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Various concentration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.