Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCLP Standard, Ricca Chemical
SDP

Catalog No. RTCLP100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RTCLP-100 100 mL
RTCLP-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RTCLP-100 Supplier Ricca Chemical Company Supplier No. RTCLP100
Add to Cart
Add to Cart

Specifications

Chemical Name or Material TCLP Standard
Packaging Amber Poly Bottle
CAS 7697-37-2, 7732-18-5, 10022-31-8, 1327-53-3, 7439-92-1
CAS Min % 91.0, 7.21, 0.09, 0.02, 0.0029, 0.0022
CAS Max % 94.0, 7.5, 0.09, 0.02, 0.0031, 0.0022
Physical Form Liquid
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.