Learn More
Invitrogen™ TCP-1 zeta Polyclonal Antibody

Description
TCP-1 zeta / CCT6A Polyclonal Antibody detects endogenous levels of TCP-1 zeta / CCT6A protein.
Specifications
Specifications
| Antigen | TCP-1 zeta |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CCT6A |
| Gene Accession No. | P40227, P80317 |
| Gene Alias | acute morphine dependence related protein 2; acute morphine dependence-related protein 2; amino acid transport defect-complementing; CCT zeta1; CCT- zeta1; CCT- zeta-1; CCT zeta-1; Cct6; Cct6a; Cctz; Cctz1; Cctz-1; CCT-zeta; CCT-zeta-1; chaperonin containing T-complex subunit 6; chaperonin containing TCP-1; chaperonin containing TCP1 subunit 6A; chaperonin containing Tcp1, subunit 6A (zeta 1); chaperonin containing Tcp1, subunit 6a (zeta); chaperonin subunit 6a (zeta); histidine transp; histidine transport regulator 3; HTR3; MoDP-2; T-complex protein 1 subunit zeta; T-complex protein 1, zeta subunit; TCP 1 zeta; TCP1 zeta; TCP-1- zeta; TCP1- zeta; TCP-1-zeta; Tcp20; TCPZ; TTCP20 |
| Gene Symbols | CCT6A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.