Learn More
Aviva Systems Biology Corp TCP10L Antibody - middle region (ARP45902_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45902_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TCP10L. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TCP10L. Alternate names: PRED77, TCP10A-2, C21orf77ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59163
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.