Learn More
Invitrogen™ TCR alpha/beta Monoclonal Antibody (TCR-3)

Description
TCR alpha/beta Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | TCR alpha/beta |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TCR-3 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | BBS with proprietary stabilizer and no preservative; pH 8.2 |
| Gene | TCRA |
| Gene Accession No. | 0 |
| Gene Alias | FLJ22602; IMD7; LOC290071; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; RGD1359684; similar to RIKEN cDNA A430107P09 gene; T cell receptor beta locus; T3/TCR complex; TCB; TCBC1; t-cell antigen receptor; T-cell receptor alpha constant; T-cell receptor beta chain; T-cell receptor V alpha; tcr alpha; TCR alpha/ beta; TCR beta; TCRA; Tcrb; TRA; Tra29; TRAC; TRB; TRB@; TRCA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.