Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR delta Monoclonal Antibody (TS-1)
GREENER_CHOICE

Catalog No. PITCR1055 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PITCR1055 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PITCR1055 Supplier Invitrogen™ Supplier No. TCR1055
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

TCR1055 targets TCR Delta TCS1 in FACS, IHC(F), IP, and TCA applications and shows reactivity with Human samples. The TCR1055 immunogen is human TCR Vdelta1-Jdelta1. TCR1055 detects TCR Delta TCS1 which has a predicted molecular weight of approximately 17 kDa.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR delta
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation
Classification Monoclonal
Clone TS-1
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA and 0.1% sodium azide
Gene TRDV1
Gene Accession No. A0A1B0GX56
Gene Alias CD3 antigen; CD3d antigen; OKT3
Gene Symbols TRD, TRDV1
Host Species Mouse
Immunogen Human TCR Vdelta1-Jdelta1.
Purification Method Purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28518, 6964
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.