Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR DO11.10 Monoclonal Antibody (KJ1-26 (KJ126, KJI-26, KJI26)), Biotin, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$140.00 - $169.00

Specifications

Antigen TCR DO11.10
Clone KJ1-26 (KJ126, KJI-26, KJI26), KJI-26, KJI26)
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-119-31
Encompass_Preferred
View Documents
Invitrogen™
13580881
50 μg
Each for $140.00
Only null left
Add to Cart
 
50-119-32
Encompass_Preferred
View Documents
Invitrogen™
13580882
100 μg
Each for $169.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: The KJ1-26 anti-idiotypic monoclonal antibody binds the T-cell receptor (TCR) expressed on DO11.10 and DO11.10.24 murine T cell hybridomas, but not on any other T cell hybridomas tested.The TCR expressed on DO11.10 recognizes an ovalbumin peptide in the context of I-A^{d^} major histocompatibility molecules.The use of DO11.10 and KJ1-26 have proven to be instrumental in describing many of the intricate details of T cell immunity. Applications Reported: This KJ1-26 (KJ126, KJI-26, KJI26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This KJ1-26 (KJ126, KJI-26, KJI26) antibody has been tested by flow cytometric analysis of the DO11.10 T cell hybridoma. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

TCR DO11 (DO11.10) is a receptor found on lymphocytes of DO11.10 transgenic mice and T-cell receptors of BALB/c-derived DO11.10 and DO11.10.24 T cell hybridomas. The DO11.10 TCR recognizes a C-terminal ovalbumin peptide (OVA 323-339) in the context of I-Ad major histocompatibility molecules. The use of antibodies against DO11.10 have proven to be instrumental in describing many of the intricate details of T cell immunity, including T cell activation, development, and function.
Specifications

Specifications

TCR DO11.10
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
D011.10; T cell receptor alpha chain; Tcra; Tcralpha
Tcra
Primary
4°C, store in dark, DO NOT FREEZE!
Tcra
KJ1-26 (KJ126, KJI-26, KJI26), KJI-26, KJI26)
Flow Cytometry
Biotin
Mouse
Mouse
P01849
21473
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.