Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR H-Y (male antigen) Monoclonal Antibody (T3.70), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5010079
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-100-79 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-100-79 Supplier Invitrogen™ Supplier No. 11993082
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The T3.70 monoclonal antibody reacts with the transgenic alpha betaTCR that recognizes the male antigen H-Y in the context of H-2D^b. A large fraction of T cells in H-Y TCR transgenic mouse expresses this receptor. The H-Y TCR transgenic mouse has been used extensively to study T cell development and the role of thymic major histocompatibility complex in CD4^+ and CD8^+ T cell differentiation. Applications Reported: The T3.70 antibody has been reported for use in flow cytometric analysis. Applications Tested: The T3.70 antibody has been tested by flow cytometric analysis of CD11.3 line, which is derived from a thymic tumor of the H-Y TCR transgenic mice and expresses the transgenic H-Y TCR. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

A large fraction of T cells in H-Y TCR transgenic mouse expresses this receptor. The H-Y TCR transgenic mouse has been used extensively to study T cell development and the role of thymic major histocompatibility complex in CD4+ and CD8+ T cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR H-Y (male antigen)
Applications Flow Cytometry
Classification Monoclonal
Clone T3.70
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias Male antigen HY; TCR HY
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.