Learn More
Description
TCR2654 targets TCR V beta 12 in FACS, IHC(F), IP, and TCA applications and shows reactivity with Human samples. The TCR2654 immunogen is human TCR V beta 12.
Specifications
Specifications
| Antigen | TCR V beta 12 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation |
| Classification | Monoclonal |
| Clone | S511 |
| Concentration | 0.15 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.5% BSA and 0.1% sodium azide |
| Gene | TRB |
| Gene Accession No. | P04435 |
| Gene Alias | T cell receptor beta chain; T cell receptor beta locus; TCRB; TCRbeta; Tib; TRB; TRB@ |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
