Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR V beta F1 Monoclonal Antibody (8A3)
GREENER_CHOICE

Catalog No. ENTCR1151
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENTCR1151 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENTCR1151 Supplier Invitrogen™ Supplier No. TCR1151
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

TCR1151 targets TCR Beta F1 in FACS, FISH, IHC, IHC(P), IP, and WB applications and shows reactivity with Human samples. The TCR1151 immunogen is human TCR β chain constant region. TCR1151 detects TCR Beta F1 which has a predicted molecular weight of approximately 13 kDa.

The T Cell Receptor alpha beta (TCRab) is a heterodimeric membrane receptor that consists of the highly variable alpha and beta chains expressed as part of a complex with the invariant CD3 proteins. TCRab participates in the activation of T-cells in response to an antigen presented in the context of MHC. Engagement of the TCRab initiates signaling cascades that ultimately result in cellular proliferation, differentiation, cytokine production, and/or activation-induced cell death. These signaling cascades regulate T-cell development, homeostasis, activation, acquisition of effector functions and apoptosis. Cells expressing TCRab constitute the vast majority of T cells in both human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V beta F1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8A3
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA and 0.1% sodium azide
Gene TRB
Gene Accession No. P0DSE2
Gene Alias T cell receptor beta locus; tcr beta f1; TCRB; TRB; TRB@
Gene Symbols TRB
Host Species Mouse
Immunogen Human TCR β chain constant region.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6957
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.