Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR V beta F1 Monoclonal Antibody (8A3), APC, eBioscience™
GREENER_CHOICE

Catalog No. 17576642
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-576-642 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-576-642 Supplier Invitrogen™ Supplier No. 17576642
  • $433.00
    $324.65 / Each of 1

    Save $108.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This 8A3 monoclonal antibody recognizes human TCR beta F1, a common framework determinant on the beta-subunit of the T cell receptor. Expression of TCR beta F1 has been described as a potential diagnostic marker indicating a favorable outcome for tumor patients with not otherwise specified Nodal Peripheral T-cell Lymphoma (PTCL-NOS). Furthermore, a lack of surface and cytoplasmic TCR beta F1 was also found in small cell variant of T-cell prolymphocytic Leukemia. This antibody will not stain the native form of the TCR and will require fixing and permeabilizing cells. Applications Reported: This 8A3 antibody has been reported for use in flow cytometric analysis, immunofluorescent staining of frozen tissue sections, and immunocytochemistry. Applications Tested: This 8A3 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

The T Cell Receptor alpha beta (TCRab) is a heterodimeric membrane receptor that consists of the highly variable alpha and beta chains expressed as part of a complex with the invariant CD3 proteins. TCRab participates in the activation of T-cells in response to an antigen presented in the context of MHC. Engagement of the TCRab initiates signaling cascades that ultimately result in cellular proliferation, differentiation, cytokine production, and/or activation-induced cell death. These signaling cascades regulate T-cell development, homeostasis, activation, acquisition of effector functions and apoptosis. Cells expressing TCRab constitute the vast majority of T cells in both human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V beta F1
Applications Flow Cytometry
Classification Monoclonal
Clone 8A3
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene TRB
Gene Accession No. P0DSE2
Gene Alias T cell receptor beta locus; tcr beta f1; TCRB; TRB; TRB@
Gene Symbols TRB
Host Species Mouse
Immunogen Human TCR beta chain constant region.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6957
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.