Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR V delta 1 Monoclonal Antibody (TS8.2)
GREENER_CHOICE

Catalog No. PITCR1730 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PITCR1730 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PITCR1730 Supplier Invitrogen™ Supplier No. TCR1730
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

TCR1730 targets TCR V delta 1 in FACS, IHC (F), and TCA applications and shows reactivity with Human samples. The TCR1730 immunogen is human TCR V delta 1.

The T Cell Receptor gamma-delta (TCR gamma delta) is a heterodimeric membrane receptor that consists of the highly variable gamma and delta chains expressed as part of a complex with the invariant CD3 proteins. Gamma-delta T cells are less frequent than alpha-beta T cells but compose a substantial part of T cells found at epithelial borders including the gut mucosa resident intraepithelial lymphocytes (IEL). The antigenic stimuli required for gamma-delta T cell receptor activation are not well understood and are thought to be mainly MHC class-independent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V delta 1
Applications Flow Cytometry, Immunohistochemistry (Frozen), T-Cell Activation
Classification Monoclonal
Clone TS8.2
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA and 0.1% sodium azide
Gene TRD
Gene Accession No. 0
Gene Symbols TRD
Host Species Mouse
Immunogen Human TCR V delta 1.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6964
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.