Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR V delta 1 Monoclonal Antibody (TS8.2), Brilliant Violet™ 421, eBioscience™
GREENER_CHOICE

Catalog No. 404567942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-456-7942 100 Tests
40-456-7941 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-456-7942 Supplier Invitrogen™ Supplier No. 404567942
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

DescriptionThis TS8.2 monoclonal antibody recognizes human TCR V delta 1, a common framework determinant on the delta-subunit of the T cell receptor. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity. TCR V delta 1 Monoclonal Antibody (TS8.2) has been validated for the following applications Flow CytometryApplications TestedThis TS8.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Blocking BuffersWhen using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) or Brilliant Stain Buffer (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer or Brilliant Stain Buffer for more information.Fixationcompensation. Some Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 μL of cell sample + 100 μL of IC Fixatio...

The T Cell Receptor gamma-delta (TCR gamma delta) is a heterodimeric membrane receptor that consists of the highly variable gamma and delta chains expressed as part of a complex with the invariant CD3 proteins. Gamma-delta T cells are less frequent than alpha-beta T cells but compose a substantial part of T cells found at epithelial borders including the gut mucosa resident intraepithelial lymphocytes (IEL). The antigenic stimuli required for gamma-delta T cell receptor activation are not well understood and are thought to be mainly MHC class-independent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V delta 1
Applications Flow Cytometry
Classification Monoclonal
Clone TS8.2
Concentration 5 μL/Test
Conjugate Brilliant Violet 421
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene TRD
Gene Symbols TRD
Host Species Mouse
Immunogen Full length protein of Human TCR delta v1.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6964
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.