Learn More
Tocris Bioscience™ TCS 3035
Description
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | TCS 3035 |
| Molecular Weight (g/mol) | 279.27 |
| Inhibitors | TCS 3035 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.