Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCTEX1D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562008 Supplier Invitrogen™ Supplier No. PA562008
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQLTKHLSEN IKDKLKEMGF DRYKMVVQVV IGEQRGEGVF MASRCFWDAD Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Dyneins are multi-subunit, high molecular weight ATPases that interact with microtubules to generate force by converting the chemical energy of ATP into the mechanical energy of movement. Cytoplasmic or axonemal Dynein is an approximately 12 subunit complex of two heavy chains, two intermediate chains to anchor Dynein to its cargo, four smaller intermediate chains and several light chains. It performs functions necessary for cell survival such as organelle transport and centrosome assembly. TCTEX1 is a cytoplasmic dynein light chain found in a complex with Na+ CP type X (SCN10A). TCTEX1D2, also called TCTEX1 domain containing 2, belongs to the dynein light chain TCTEX-type family. Its function is still under investigation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTEX1D2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCTEX1D2
Gene Accession No. Q8WW35
Gene Alias 0610012D17Rik; 2410011K15Rik; dynein light chain Tctex-type 2B; DYNLT2B; RGD1563597; Tctex1 domain containing 2; tctex1 domain-containing protein 2; TCTEX1D2; Unknown (protein for IMAGE:8040601)
Gene Symbols TCTEX1D2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCTEX1D2. Recombinant protein control fragment (Product #RP-100327).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255758
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.