Learn More
Pharma Alliance Group TD-02 TAP DENSITY TESTER
Supplier:  Pharma Alliance Group TD02
Encompass
Supports USP 1 (300 taps per min) and USP 2 (250 taps per min) and is also available ASTM test method - Simultaneous rotating and tapping motion assures an evenly packed surface - Calculation of test results like tapped density, compressibility index and Hausner ratio - Printer port for documenting test results as per GMP/GLP standard - LAN connectivity for data transfer to a computer. Tap Height: USP I : 14 mm /- 2 mm, USP II : 3 mm /- 0.2 mm. Tap/MinuteUSP I : 300 mm /- 15, USP II : 250 /- 15. Display:20 x 4 character LCD. Dimensions: 36 cm x 20 cm x 32 cm
Catalog No. NC0433377
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.