Learn More
Invitrogen™ TDE1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total SERINC3 protein.
Specifications
Specifications
| Antigen | TDE1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Serinc3 |
| Gene Accession No. | Q13530, Q9QZI9 |
| Gene Alias | AA959945; AA960047; Aigp1; AIGP-1; axotomy-induced glyco/Golgi protein 1; Axotomy-induced glycoprotein 1; DIFF33; Membrane protein TMS-1; SBBI99; Serinc3; Serine incorporator 3; TDE; TDE1; Tms1; TMS-1; tumor differentially expressed 1; tumor differentially expressed protein 1 |
| Gene Symbols | Serinc3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.