Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TDO2 Monoclonal Antibody (OTI4G2)
GREENER_CHOICE

Catalog No. PIMA525965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525965 Supplier Invitrogen™ Supplier No. MA525965
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TDO2 Monoclonal Antibody for Western Blot, IHC (P)

Tryptophan 2,3-dioxygenase (TDO2) is an enzyme encoded by the TDO2 gene, predominantly expressed in the liver and playing a crucial role in tryptophan metabolism through the kynurenine pathway. TDO2 catalyzes the first and rate-limiting step of this pathway, converting tryptophan into N-formylkynurenine. This metabolic process is vital for maintaining systemic tryptophan homeostasis and regulating its availability for protein synthesis and serotonin production. Beyond its metabolic function, TDO2 is involved in modulating immune responses. It contributes to immune tolerance by depleting tryptophan, thereby inhibiting T cell proliferation and promoting regulatory T cell development. Recent research has identified TDO2's role in cancer, particularly in shaping the tumor microenvironment and facilitating immune escape mechanisms through tryptophan catabolism. Its expression is upregulated in several cancers, including gliomas, correlating with tumor severity and patient prognosis. TDO2 and its associated pathways have become therapeutic targets in cancer immunotherapy due to their influence on immune system modulation and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDO2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TDO2
Gene Accession No. P48775
Gene Alias AA407491; chky; HAMAP-Rule:MF_03020}; TDO; TDO {ECO:0000255; tdo variant1; tdo variant2; TDO2; TO; TO {ECO:0000255; TPH2; TRPO; TRPO {ECO:0000255; tryptamin 2,3-dioxygenase; tryptamin 2,3-dioxygenase {ECO:0000255; Tryptophan 2,3-dioxygenase; tryptophan 2,3-dioxygenase {ECO:0000255; tryptophan 2,3-dioxygenase A; tryptophan 2,3-dioxygenase A; tryptophan 2,3-dioxygenase; tryptophan oxygenase; tryptophan oxygenase {ECO:0000255; Tryptophan pyrrolase; tryptophan pyrrolase {ECO:0000255; tryptophan-2,3-dioxygenase; tryptophan-23-dioxygenase; tryptophanase; tryptophanase {ECO:0000255
Gene Symbols TDO2
Host Species Mouse
Immunogen Full length human recombinant protein of TDO2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6999
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.