Learn More
Invitrogen™ TDP-43 Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-20408 can be used with blocking peptide PEP-0525. The immunogen is located within amino acids 60 - 110 of TDP43.
Specifications
Specifications
| Antigen | TDP-43 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | TARDBP |
| Gene Accession No. | Q13148, Q921F2 |
| Gene Alias | 1190002A23Rik; ALS10; C85084; hypothetical protein; OTTHUMP00000002171; OTTHUMP00000002173; RCJMB04_5g9; RP4-635E18.2; TAR DNA binding protein; TAR DNA binding protein L homeolog; TAR DNA-binding protein 43; TAR DNA-binding protein-43; tardbp; tardbp.L; Tdp43; TDP-43; wu:fb77f02; wu:fc52g10; XELAEV_18035188mg |
| Gene Symbols | TARDBP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.