Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDRD12 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA571265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571265 Supplier Thermo Scientific Supplier No. PA571265

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Horse: 86%; Human: 100%; Pig: 86%

TDRD12 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TDRD12
Gene Accession No. Q587J7
Gene Alias ECAT8; ES cell associated transcript 8; ES cell-associated transcript 8 protein; Putative ATP-dependent RNA helicase TDRD12; TDRD12; tudor domain containing 12; tudor domain-containing protein 12
Gene Symbols TDRD12
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human TDRD12
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91646
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.