Learn More
MilliporeSigma™ Supelco™ TDS3™ Storage Container
Catalog No. 111011855 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Dynatherm 850/890, ACEM 900/901-FF (Standard Sampling Tube)
Gerstel TDS 2/TDS/A, Tekmar AEROtrap 6000
Gerstel™ 60 mm TDU Tubes
PerkinElmer ATD-50/ATD-400/Turbomatrix, Markes International Unity
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-101-1855 Gerstel™ 60 mm TDU Tubes
11-101-1745 Dynatherm 850/890, ACEM 900/901-FF (Standard Sampling Tube)
11-101-1735 Gerstel TDS 2/TDS/A, Tekmar AEROtrap 6000
11-101-1736 PerkinElmer ATD-50/ATD-400/Turbomatrix, Markes International Unity
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-101-1855 Supplier MilliporeSigma™ Supelco™ Supplier No. 28307U
Add to Cart
Add to Cart
Simplifies the Sample Preparation Process for Thermal Desorption
  • Containers help to maintain the sample integrity of both the tubes waiting to be sampled, or those which already have a sample adsorbed
TRUSTED_SUSTAINABILITY
Specifications
Product Type TDS3™ Storage Container
For Use With (Equipment) Gerstel™ 60 mm TDU Tubes
For Use With (Application) Maintaining Sample Integrity
Quantity 1 Ea.
TDS3 is a trademark of Sigma-Aldrich Co. LLC