Learn More
R&D Systems™ TdT In Situ Apoptosis Detection Kit - Core Reagents
SDP
Catalog No. 481030CK
Change view
Click to view available options
Quantity:
30 Tests
Catalog No. Quantity
481030CK 30 Tests
1 options
This item is not returnable. View return policy
Catalog No. 481030CK Supplier R&D Systems™ Supplier No. 481030CK
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
10X TdT Labeling Buffer, 100mL; 10X TdT Stop Buffer, 100mL; TdT dNTP Mix, 30uL; TdT Enzyme, 30uL; Strep-HRP, 30uL; 30uL each of 50X Co+2, 50X Mg+2, 50X Mn+2
Restocks the key components of TACS kits
Core reagents for the TACS kits include: 10X TdT Labeling Buffer, 100mL; 10X TdT Stop Buffer, 100mL; TdT dNTP Mix, 30uL; TdT Enzyme, 30uL; Strep-HRP, 30uL
Also includes 30uL each of 50X Co+2, 50X Mg+2, 50X Mn<special
Specifications
Quantity 30 Tests
Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.