Learn More
R&D Systems™ TdT In Situ Apoptosis Detection Kit - Replenisher
SDP
Catalog No. 481030R
Change view
Click to view available options
Quantity:
30 Tests
Catalog No. Quantity
481030R 30 Tests
1 options
This item is not returnable. View return policy
Catalog No. 481030R Supplier R&D Systems™ Supplier No. 481030R
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
Proteinase K Solution, 30uL; TdT dNTP Mix, 30uL; TdT Enzyme, 30uL; Strep-HRP, 30uL; 30uL each of 50X cations
Provides only reagents most rapidly exhausted from the complete TACS 2 TdT kits
The replenisher is also useful when using alternative detection and counterstaining methods in double-labeling experiments
Abbreviated instructions compared to the complete TACS 2 kits
Specifications
Quantity 30 Tests
Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.