Learn More
Biocare Medical, LLC Terminal Deoxynucleotidyl Transferase Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CP134AK
Encompass_Preferred
Terminal Deoxynucleotidyl Transferase antibody (TdT) is a nuclear protein widely used as a marker for lymphoblastic leukemia. TdT is a template-independent DNA polymerase which has been shown to be responsible for the addition of nucleotides at the N-region junction of rearranged Ig heavy chain and T-cell receptor gene segments during the maturation of B- and T-cells. Studies have shown that a panel of antibodies consisting of TdT, CD10, CD99 (MIC2), Bcl-2 and CD34 can be used to distinguish lymphoblastic leukemias from small noncleaved cell lymphomas.
Catalog No. 50-827-63
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.