Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TE buffer, (20X, pH 7.6), RNAse free

Catalog No. AAJ62285AK
Change view
Click to view available options
Quantity:
125 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62285AK 250 mL
AAJ62285EQE 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62285AK Supplier Thermo Scientific Chemicals Supplier No. J62285AK
Add to Cart

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

TE buffer is a commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

Solubility
Fully miscible in water.

Notes
200mM Tris-HCl with 20mM EDTA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00236359
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

pH 7.6
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully miscible in water.
Chemical Name or Material TE buffer, RNAse free
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.