Learn More
Therapak™ TearTech™ Tube Shuttles
Catalog No. 22130038 Shop All Therapak Corporation Products
Change view
Click to view available options
Closure Type:
TearTech
Zipper
Quantity:
Up to 6 tubes
Up to 8 tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Quantity
22-130-038 Zipper Up to 8 tubes
22-130-019 TearTech Up to 6 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-130-038 Supplier Therapak™ Supplier No. 15540
Add to Cart
Add to Cart
Separates tubes to prevent contact
  • Leak-tight construction for increased safety during transport
  • Numbered tube slots for easy reference
  • Two sizes available to hold six or eight blood or urine tubes (12–16mm dia.)
  • Six-tube shuttle features a TearTech closure that folds down for a permanent seal, yet provides a natural perforation for easy opening at the lab
  • Eight tube shuttle features a zip-style closure
  • Shuttles can be rolled or folded and placed inside a secondary watertight container
Specifications
Quantity Up to 8 tubes
Applications Blood and urine tube mailings
Closure Type Zipper