Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tebuconazole

Catalog No. AC463260050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463260050 5 g
AC463260010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463260050 Supplier Thermo Scientific Chemicals Supplier No. 463260050

CAS: 107534-96-3 | C16H22ClN3O | 307.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107534-96-3
Molecular Formula C16H22ClN3O
Molecular Weight (g/mol) 307.82
MDL Number MFCD02674797
InChI Key PXMNMQRDXWABCY-UHFFFAOYNA-N
IUPAC Name 1-(4-chlorophenyl)-4,4-dimethyl-3-[(1H-1,2,4-triazol-1-yl)methyl]pentan-3-ol
SMILES CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN1C=NC=N1

Specifications

Melting Point 102.0°C to 105.0°C
Flash Point >100°C
Quantity 5 g
Formula Weight 307.82
Chemical Name or Material Tebuconazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Suspected of damaging the unborn child.

Very toxic to aquatic life.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.