Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex TECAN Extractor Rack Stabilizer
SDP

Catalog No. 22000562
Change view
Click to view available options
Positions:
1
2
3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
22-000-562 2
22-000-561 1
22-000-563 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-562 Supplier Poltex Supplier No. TECANRK2
Add to Cart
Add to Cart

Holds and stabilizes sample tube racks for Tecan or other related extraction/processing machines.

  • Comes in 1, 2, or 3 rack capacity
  • Super durable and cleanable (alcohol resistant) PETG construction

Specifications

Autoclavable Not Autoclavable
Color Clear
Material Nylon-coated Stainless Steel
Positions 2
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Extractor Racks
Size Small
Product Type Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.