Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Utility Pants
SDP

Catalog No. 17355195 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
30 x 34 in.
32 x 34 in.
34 x 34 in.
36 x 34 in.
38 x 34 in.
40 x 34 in.
42 x 34 in.
44 x 34 in.
46 x 34 in.
48 x 34 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-195 44 x 34 in.
NAN 30 x 34 in.
17-355-177 32 x 34 in.
17-355-180 34 x 34 in.
17-355-183 36 x 34 in.
17-355-186 38 x 34 in.
17-355-189 40 x 34 in.
17-355-192 42 x 34 in.
17-355-198 46 x 34 in.
17-355-201 48 x 34 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 17-355-195 Supplier National Safety Apparel™ Supplier No. PNT9SU44X34
Add to Cart
Add to Cart
This item is not returnable. View return policy

FR utility pants offer durable construction and high abrasion resistance.

  • Reliable Dual Hazard Protection: Provides safety against arc flash and flash fire for peace of mind on the job
  • Heavy-Duty Performance: Built for ultimate durability to withstand tough work environments
  • High Abrasion Resistance: Made with rugged construction to handle wear and tear
  • Ample Storage Options: Left-side cargo pocket with flap, plus a double tool pocket on the right to hold a variety of tools
  • Classic Utility Pockets: Standard hip and back pockets for everyday essentials

Specifications

Size 44 x 34 in.
Gender Unisex
Product Type Flame-resistant Work Pants
Material FR Cotton
Color Navy
Seam Type Stitched
No. of Pockets 7
Closure Type Zipper, Button
Disposable Reusable
Hazard Risk Category CAT 2
Arc Flash Rating 14 cal/cm²
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
Size (English) Inseam 34 in.
Size (English) Waist 44 in.
For Use With (Application) Utilities, Oil and Gas
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.