Learn More
Getinge USA Tec Wash III, 5G
Supplier:  Getinge USA 61301667776
Encompass
Tec Wash III is an aluminum safe, alkaline detergent designed for use in a wide variety of operating conditions. For use in mechanical cleaning equipment, cart washers, instrument washers and washer-disinfectors.
Catalog No. NC0981900
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.