Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Tecnu™ Oak-n-Ivy Cleansing Treatment

Catalog No. 19090855 Shop All Medique Products
Change view
Click to view available options
Quantity:
4 oz Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-855 4 oz Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-855 Supplier Medique Supplier No. 8360
Add to Cart
Add to Cart

Removes toxic oils of poison oak, ivy and sumac.

  • Non-irritating, non-sticky lotion

Specifications

Product Type Oak-n-Ivy Cleansing Treatment
For Use With (Application) Removes toxic oils of poison oak, ivy, and sumac
Format Non-irritating, Non-sticky Lotion
Packaging Individually Packaged
Quantity 4 oz Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.