Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tecrl Monoclonal Antibody (601CT22.4.3)
GREENER_CHOICE

Catalog No. PIMA537597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537597 Supplier Invitrogen™ Supplier No. MA537597
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tecrl Monoclonal Antibody for Western Blot

The protein encoded by this gene contains a ubiquitin-like domain in the N-terminal region, three transmembrane segments and a C-terminal 3-oxo-5-alpha steroid 4-dehydrogenase domain. The protein belongs to the steroid 5-alpha reductase family. Mutations in this gene result in ventricular tachycardia, catecholaminergic polymorphic, 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tecrl
Applications Western Blot
Classification Monoclonal
Clone 601CT22.4.3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.09% sodium azide; pH 7.4
Gene TECRL
Gene Accession No. Q5HYJ1
Gene Alias D330017N19Rik; glycoprotein, synaptic 2-like; GPSN2L; SRD5A2L2; steroid 5 alpha-reductase 2-like 2; steroid 5-alpha-reductase 2-like 2 protein; synaptic glycoprotein SC2-like; TECRL; TERL; trans-2,3-enoyl-CoA reductase like; trans-2,3-enoyl-CoA reductase-like; Unknown (protein for MGC:128084)
Gene Symbols TECRL
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 325-354 amino acids from the C-terminal region of human SRD5A2L2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.