Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Tectonics Model Set
SDPSCI_ED

Catalog No. S27859 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Replacement Sand
Tectonics Model Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S27859 Replacement Sand
S27858 Tectonics Model Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27859 Supplier United Scientific Supplies Supplier No. TCTN01SAND
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demonstrates plate tectonic principles, plate boundary interactions, and the geometry/relative motions of faulting of geological layers

  • Contains a Fault 3D Model Set (5 Manipulatives) and a Tectonics Sand Layer Model
  • Fault Model Set demonstrates normal faulting (extension), reverse faulting (compression), and horizontal slip faulting (shear)
  • Tectonics Sand Layer Model explores orogeny of the Appalachian Mountains, including coal formation in the sedimentary Appalachian Basin and its coal seam
  • Construct a sand layer model system mimicking a subduction zone of oceanic crust (Juan de Fuca Plate)
  • Simulate the creation of the San Andreas Fault
  • Includes three hands-on, geoinformation activities for students to work on using metadata (Map the Continental Shelf, Tectonic Plate Movement, Earthquake Prediction)
  • Access code is provided to download interactive Teacher and Student Guides, videos, geoinformation data sets, and other resource information

Specifications

For Use With (Application) Demonstration of Plate Tectonics
Product Type Replacement Sand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.