Learn More
Grainger Tee Forged Steel 1/2 in Pipe Size FNPT
Supplier:  Grainger 14J734
Encompass_Preferred
The Grainger 1/2" Forged Steel Tee is made from forged steel for durability and has 1/2" female NPT threaded connections. It is rated for a maximum pressure of 3000 psi at 100°F and can be used for air, oil, natural gas, and steam applications. The tee provides a branch flow path from a main pipe run.
Catalog No. 50-275-0607
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.