Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEF4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988368
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8368 Supplier Bioss Supplier No. BS8212R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5′-GTGGAATGT-3′). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.

Specifications

Antigen TEF4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TEAD2
Gene Accession No. Q15562
Gene Alias BOS_17592; Embryonic TEA domain containing factor; Embryonic TEA domain-containing factor; Etdf; ETEF-1; Etf; LOC100352383; TEA domain family member 2; TEA domain family member 2-like; TEA domain transcription factor 2; Tead2; TEAD-2; Tef4; TEF-4; Transcriptional enhancer factor TEF-4
Gene Symbols TEAD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TEA domain family member 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8463
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.