Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Teflon™ Bushing
GREENER_CHOICESDP

Catalog No. 31502262 Shop All Kemtech Products
Change view
Click to view available options
Diameter (Metric):
15 mm
25 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
31-502-262 15 mm
31-502-263 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-502-262 Supplier Synthware™ Supplier No. B100015
Add to Cart
Add to Cart

Teflon™ Bushing includes an EP™ O-ring, for use with pressure valves.

  • For use with pressure valves
  • Includes an EP™ O-ring

Specifications

Shape Rounded Grip, Screw Stem
Diameter (Metric) 15 mm
Sterility Non-sterile
For Use With (Equipment) Pressure Valves
Autoclavable Not Autoclavable
Color White
Material PTFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.