Learn More
Cable Ties And More TEFZEL CBL TIES11IN/3IN 50PCS
SDP
Supplier:  Cable Ties And More TF11120
Encompass
Material- Ethylene-tetrafluoroethylene Highly Resistant To Concentrated AcidsGood Radiation Resistance Up To 100 megaradsRoHS CompliantWeather ResistantUV Resistant - For indoor or outdoor useMax Operation Temp 302 degrees F / 150 degrees CMeet the requirements of IEEE 383
Catalog No. NC2481350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.