Learn More
Cytiva TEFZEL TUBING,2M/ID0.75/OD1/16
Supplier:  Cytiva 18111974
Encompass
EFTE tubing for use with KTA systems. White/clear tubing for easy identification. 2 m length with ID 0.75mm and OD 1/16 in.0
Catalog No. 50-105-5657