Learn More
Medchemexpress LLC HY-10080 5mg Medchemexpress, Teglarinad (chloride) CAS:432037-57-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100805MG
Encompass_Preferred
Medchemexpress, HY-10080 5mg Teglarinad (chloride) CAS:432037-57-5 Teglarinad chloride (GMX1777) is a prodrug of GMX1778 (a nicotinamide phosphoribosyl transferase inhibitor). Teglarinad chloride exhibits antitumor activity in mice can be attributed to inhibition of NAMPT. Teglarinad chloride also enhances radiation efficacy, mediated by interference with DNA repair and antiangiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0603
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.