Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Teicoplanin, cont. 80+% A2

Catalog No. AAJ66785MC
Change view
Click to view available options
Quantity:
100mg
500mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66785MC 100mg
AAJ66785ME 500mg
AAJ6678503 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66785MC Supplier Thermo Scientific Chemicals Supplier No. J66785MC
Add to Cart

CAS: 61036-62-21879.67 g/mol

A glycopeptide antibiotic produced by Actinoplanes teichomiceticus; effective against Gram-positive bacteria that are resistant to β-lactam antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61036-62-2

Specifications

Purity 80%
Content And Storage Keep cold
Format Powder
Molecular Weight (g/mol) 1879.67
Product Type Teicoplanin
Quantity 100mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.