Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Teicoplanin Recombinant Mouse Monoclonal Antibody (2F4)

Catalog No. MA566292
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566292 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566292 Supplier Invitrogen™ Supplier No. MA566292
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Recombinant Monoclonal Antibody

Teicoplanin Recombinant Monoclonal Antibody for ELISA

Teicoplanin is a glycopeptide antibiotic produced by the actinobacterium Actinoplanes teichomyceticus, primarily used to treat severe infections caused by multidrug-resistant Gram-positive pathogens. Structurally, teicoplanin consists of a complex of five closely related glycopeptides sharing a heptapeptide base and an aglycone with aromatic amino acids, d-mannose, and N-acetyl-d-glucosamine, with a molecular weight ranging from 1562 to 1891 Da. It is structurally similar to vancomycin and functions by inhibiting bacterial cell wall synthesis, making it effective against various Gram-positive bacteria. Teicoplanin has been explored for its potential use in treating COVID-19 due to its ability to inhibit the viral life cycle of coronaviruses, including SARS-CoV-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Teicoplanin
Applications ELISA
Classification Recombinant Monoclonal
Clone 2F4
Concentration 7.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen Conjugate of TCL hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.