Learn More
Celestron™ Telescope, Microscrope, and Binocular Science Kit
SCI_ED
Catalog No. S81330 Shop All Celestron International Products
Change view
Click to view available options
Magnification Power:
900X
Catalog No. Magnification Power
S81330 900X
1 options
Warning: Contains Chemicals Listed as Cancer and Reproductive Harm in the State of California
Catalog No. S81330 Supplier Celestron™ Supplier No. 22010
Add to Cart
Add to Cart
Warning: Contains Chemicals Listed as Cancer and Reproductive Harm in the State of California
Celestron™ Telescope, Microscrope, and Binocular Science Kit is perfect for budding scientists
  • Star gaze with the large 50 mm telescope that includes a fill height adjustable tripod, a finderscope for easily lining up your sight, and two powerful eyepieces
  • View the craters of the moon, explore your favorite constellations and even witness the rings of Saturn
  • The illuminated microscope magnifies up to 900x with additional powers of 150x and 450x and includes blank and prepared slides, cover slips, blank labels, spatula, dissecting probe, eye dropper, tweezers, spare light bulbs and specimen vials
  • The powerful binoculars are great for exploring the amazing landscapes, animals, and astonishing occurrences that are happening all around
  • Featuring 4x magnification and a 30 mm coated glass ive lenses, 150x/450x/900x magnification (2 AA batteries - Not Included)
  • Microscope accessories include 3 blank microscope slides, 3 cover slips, 3 blank labels, spatula, dissecting probe, plastic scalpel, eyedropper, tweezers, 3 specimen vials, spare light bulb
  • Telescope accessories include adjustable tripod, 2 eyepieces (5 mm, 10 mm), finderscope, diagonal
  • All 3 devices built strong with professional craftsmanship for exceptional viewing experience
  • This complete optical science kit combines a 50 mm refractor telescope with a complete microscope kit and 4x binoculars
  • Suitable for ages 10 years and above
Specifications
Lighting Type LED
Eyepiece Fixed
Head Style Monocular
Product Type Science Kit
Includes 50 mm Refractor Telescope, Complete Microscope Kit, 4x Binoculars
Magnification Power 900X
Objective Types Achromatic
For Use With (Application) Ages 10+