Learn More
Corning™ Telescopic Suction Tubes
Catalog No. 07201134 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 to 10 mL
25 to 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07-201-134 2.5 to 10 mL
07-201-135 25 to 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-201-134 Supplier Corning™ Supplier No. 6867
Add to Cart
Add to Cart
Corning™ Telescopic Suction Tubes for quick and precise dispensing of acids, bases and solvents
  • Suction tube
  • For use with Bottle Top Dispenser
TRUSTED_SUSTAINABILITY
Specifications
Product Type Telescopic Suction Tube
Autoclavable Autoclavable
Certifications/Compliance ISO 8655
Volume (Metric) 2.5 to 10 mL
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) 2.5-10 mL Bottle Top Dispenser
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov